Ethyl-5-oxo-5-(2-pyridyl)valerate ≥97%

Supplier: Matrix Scientific

087102-1G 087102-2G 087102-5G
102852-508EA 490.77 USD
102852-508 102852-510 102852-512
Ethyl-5-oxo-5-(2-pyridyl)valerate ≥97%
Ethyl-5-oxo-5-(2-pyridyl)valerate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD01320355
CAS Number: 898776-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR