Ethyl-6-oxononanoate ≥97%

Supplier: Matrix Scientific

087095-1G 087095-2G 087095-5G
102852-466EA 592.31 USD
102852-466 102852-468 102852-470
Ethyl-6-oxononanoate ≥97%
Ethyl-6-oxononanoate

Hazard: Irritant

Formula: C₁₁H₂₀O₃
MDL Number: MFCD01320349
CAS Number: 4144-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR