Ethyl-6-oxodecanoate ≥97%

Supplier: Matrix Scientific

087089-1G 087089-2G 087089-5G
102852-430EA 592.31 USD
102852-430 102852-432 102852-434
Ethyl-6-oxodecanoate ≥97%
Ethyl-6-oxodecanoate

Hazard: Irritant

Formula: C₁₂H₂₂O₃
MDL Number: MFCD01320342
CAS Number: 4144-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR