Ethyl-5-oxononanoate ≥97%
Supplier: Matrix Scientific
087088-1G
087088-2G
087088-5G
102852-424EA
592.31
USD
102852-424
102852-426
102852-428
Ethyl-5-oxononanoate ≥97%
Ethyl-5-oxononanoate
Hazard: Irritant
Formula:
C₁₁H₂₀O₃ |
MDL Number:
MFCD01320341 CAS Number: 24071-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...