Ethyl-4-oxooctanoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-oxooctanoate
087087-1G
087087-2G
087087-5G
102852-418EA
592.31
USD
102852-418
102852-420
102852-422
Ethyl-4-oxooctanoate ≥97%
Ethyl-4-oxooctanoate
Hazard: Irritant
Formula:
C₁₀H₁₈O₃ MW: 186.25 g/mol |
MDL Number:
MFCD00520679 CAS Number: 37174-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...