Ethyl-8-oxodecanoate ≥97%

Supplier: Matrix Scientific

087086-1G 087086-2G 087086-5G
102852-412EA 592.31 USD
102852-412 102852-414 102852-416
Ethyl-8-oxodecanoate ≥97%
Ethyl-8-oxodecanoate

Hazard: Irritant

Formula: C₁₂H₂₂O₃
MDL Number: MFCD01320340
CAS Number: 105906-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR