Ethyl-6-oxooctanoate ≥97%

Supplier: Matrix Scientific

087084-1G 087084-2G 087084-5G
102852-400EA 592.31 USD
102852-400 102852-402 102852-404
Ethyl-6-oxooctanoate ≥97%
Ethyl-6-oxooctanoate

Hazard: Irritant

Formula: C₁₀H₁₈O₃
MDL Number: MFCD01320339
CAS Number: 4233-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR