Ethyl-5-oxoheptanoate ≥97%
Supplier: Matrix Scientific
087083-1G
087083-2G
087083-5G
102852-394EA
986.15
USD
102852-394
102852-396
102852-398
Ethyl-5-oxoheptanoate ≥97%
Ethyl-5-oxoheptanoate
Hazard: Irritant
Formula:
C₉H₁₆O₃ |
MDL Number:
MFCD01320338 CAS Number: 70432-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...