Ethyl-4-oxohexanoate ≥97%

Supplier: Matrix Scientific

087082-1G 087082-2G 087082-5G
102852-388EA 886.15 USD
102852-388 102852-390 102852-392
Ethyl-4-oxohexanoate ≥97%
Ethyl-4-oxohexanoate

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD00068986
CAS Number: 3249-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR