Ethyl-7,7-dimethyl-6-oxooctanoate ≥95%

Supplier: Matrix Scientific

087081-1G 087081-2G 087081-5G
102852-382EA 592.31 USD
102852-382 102852-384 102852-386
Ethyl-7,7-dimethyl-6-oxooctanoate ≥95%
Ethyl-7,7-dimethyl-6-oxooctanoate

Hazard: Irritant

Formula: C₁₂H₂₂O₃
MDL Number: MFCD01320336
CAS Number: 898776-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR