Ethyl-6,6-dimethyl-5-oxoheptanoate ≥95%

Supplier: Matrix Scientific

087080-1G 087080-2G 087080-5G
102852-376EA 735.38 USD
102852-376 102852-378 102852-380
Ethyl-6,6-dimethyl-5-oxoheptanoate ≥95%
Ethyl-6,6-dimethyl-5-oxoheptanoate

Hazard: Irritant

Formula: C₁₁H₂₀O₃
MDL Number: MFCD01320335
CAS Number: 569685-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR