Ethyl-5,5-dimethyl-4-oxohexanoate ≥97%

Supplier: Matrix Scientific

087079-1G 087079-2G 087079-5G
102852-370EA 592.31 USD
102852-370 102852-372 102852-374
Ethyl-5,5-dimethyl-4-oxohexanoate ≥97%
Ethyl-5,5-dimethyl-4-oxohexanoate

Hazard: Irritant

Formula: C₁₀H₁₈O₃
MDL Number: MFCD01320334
CAS Number: 37174-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR