Ethyl-6-cyclopropyl-6-oxohexanoate ≥97%

Supplier: Matrix Scientific

087077-1G 087077-2G 087077-5G
102852-358EA 592.31 USD
102852-358 102852-360 102852-362
Ethyl-6-cyclopropyl-6-oxohexanoate ≥97%
Ethyl-6-cyclopropyl-6-oxohexanoate

Hazard: Irritant

Formula: C₁₁H₁₈O₃
MDL Number: MFCD01320332
CAS Number: 898776-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR