Ethyl-5-cyclopropyl-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087076-1G 087076-2G 087076-5G
102852-352EA 592.31 USD
102852-352 102852-354 102852-356
Ethyl-5-cyclopropyl-5-oxovalerate ≥97%
Ethyl-5-cyclopropyl-5-oxovalerate

Hazard: Irritant

Formula: C₁₀H₁₆O₃
MDL Number: MFCD01320331
CAS Number: 898776-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR