Ethyl-4-cyclopropyl-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

087075-1G 087075-2G 087075-5G
102852-346EA 590.77 USD
102852-346 102852-348 102852-350
Ethyl-4-cyclopropyl-4-oxobutyrate ≥97%
Ethyl-4-cyclopropyl-4-oxobutyrate

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD01320330
CAS Number: 184297-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR