Ethyl-6-cyclobutyl-6-oxohexanoate ≥97%

Supplier: Matrix Scientific

087074-1G 087074-2G 087074-5G
102852-340EA 592.31 USD
102852-340 102852-342 102852-344
Ethyl-6-cyclobutyl-6-oxohexanoate ≥97%
Ethyl-6-cyclobutyl-6-oxohexanoate

Hazard: Irritant

Formula: C₁₂H₂₀O₃
MDL Number: MFCD01320328
CAS Number: 898776-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR