Ethyl-5-cyclobutyl-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087073-1G 087073-2G 087073-5G
102852-334EA 592.31 USD
102852-334 102852-336 102852-338
Ethyl-5-cyclobutyl-5-oxovalerate ≥97%
Ethyl-5-cyclobutyl-5-oxovalerate

Hazard: Irritant

Formula: C₁₁H₁₈O₃
MDL Number: MFCD01320327
CAS Number: 898776-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR