Ethyl-6-cyclohexyl-6-oxohexanoate ≥97%

Supplier: Matrix Scientific

087070-1G 087070-2G 087070-5G
102852-316EA 592.31 USD
102852-316 102852-318 102852-320
Ethyl-6-cyclohexyl-6-oxohexanoate ≥97%
Ethyl-6-cyclohexyl-6-oxohexanoate

Hazard: Irritant

Formula: C₁₄H₂₄O₃
MDL Number: MFCD01320320
CAS Number: 16076-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR