Ethyl-5-cyclohexyl-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087069-1G 087069-2G 087069-5G
102852-310EA 592.31 USD
102852-310 102852-312 102852-314
Ethyl-5-cyclohexyl-5-oxovalerate ≥97%
Ethyl-5-cyclohexyl-5-oxovalerate

Hazard: Irritant

Formula: C₁₃H₂₂O₃
MDL Number: MFCD01320319
CAS Number: 16076-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR