Ethyl-4-cyclohexyl-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

087068-1G 087068-2G 087068-5G
102852-304EA 886.15 USD
102852-304 102852-306 102852-308
Ethyl-4-cyclohexyl-4-oxobutyrate ≥97%
Ethyl-4-cyclohexyl-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₂₀O₃
MDL Number: MFCD01320318
CAS Number: 54966-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR