Ethyl-4-(2-fluorophenyl)-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

087066-1G 087066-2G 087066-5G
102852-292EA 886.15 USD
102852-292 102852-294 102852-296
Ethyl-4-(2-fluorophenyl)-4-oxobutyrate ≥97%
Ethyl-4-(2-fluorophenyl)-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₁₃FO₃
MW: 224.23 g/mol
MDL Number: MFCD01320293
CAS Number: 898753-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR