Ethyl-4-(2-naphthyl)-4-oxobutyrate ≥97%
Supplier: Matrix Scientific
087064-1G
087064-2G
087064-5G
102852-280EA
490.77
USD
102852-280
102852-282
102852-284
Ethyl-4-(2-naphthyl)-4-oxobutyrate ≥97%
Ethyl-4-(2-naphthyl)-4-oxobutyrate
Hazard: Irritant
Formula:
C₁₆H₁₆O₃ MW: 256.3 g/mol |
MDL Number:
MFCD01320266 CAS Number: 25370-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...