Ethyl-5-(2-methylphenyl)-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087062-1G 087062-2G 087062-5G
102852-268EA 490.77 USD
102852-268 102852-270 102852-272
Ethyl-5-(2-methylphenyl)-5-oxovalerate ≥97%
Ethyl-5-(2-methylphenyl)-5-oxovalerate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD01320209
CAS Number: 100972-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR