Ethyl-4-(4-chlorophenyl)-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(4-Chloro-phenyl)-4-oxo-butyric acid ethyl ester

087060-1G 087060-2G 087060-5G
102852-256EA 490.77 USD
102852-256 102852-258 102852-260
Ethyl-4-(4-chlorophenyl)-4-oxobutyrate ≥97%
Ethyl-4-(4-chlorophenyl)-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₁₃ClO₃
MW: 240.69 g/mol
MDL Number: MFCD00032767
CAS Number: 53503-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR