Ethyl-7-(4-fluorophenyl)-7-oxoheptanoate ≥97%

Supplier: Matrix Scientific

087059-1G 087059-2G 087059-5G
102852-250EA 490.77 USD
102852-250 102852-252 102852-254
Ethyl-7-(4-fluorophenyl)-7-oxoheptanoate ≥97%
Ethyl-7-(4-fluorophenyl)-7-oxoheptanoate

Hazard: Irritant

Formula: C₁₅H₁₉FO₃
MDL Number: MFCD01311595
CAS Number: 122115-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR