Ethyl-4-(3-bromophenyl)-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

087057-1G 087057-2G 087057-5G
102852-238EA 490.77 USD
102852-238 102852-240 102852-242
Ethyl-4-(3-bromophenyl)-4-oxobutyrate ≥97%
Ethyl-4-(3-bromophenyl)-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₁₃BrO₃
MDL Number: MFCD00672064
CAS Number: 147374-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR