Ethyl-7-(4-bromophenyl)-7-oxoheptanoate ≥97%
Supplier: Matrix Scientific
087056-1G
087056-2G
087056-5G
102852-232EA
769.23
USD
102852-232
102852-234
102852-236
Ethyl-7-(4-bromophenyl)-7-oxoheptanoate ≥97%
Hazard: Irritant
Formula:
C₁₅H₁₉BrO₃ |
MDL Number:
MFCD00672071 CAS Number: 122115-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...