Ethyl-5-(4-bromophenyl)-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087055-1G 087055-2G 087055-5G
102852-226EA 490.77 USD
102852-226 102852-228 102852-230
Ethyl-5-(4-bromophenyl)-5-oxovalerate ≥97%
Ethyl-5-(4-bromophenyl)-5-oxovalerate

Hazard: Irritant

Formula: C₁₃H₁₅BrO₃
MDL Number: MFCD00672069
CAS Number: 898792-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR