Ethyl-4-(4-bromophenyl)-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(4-bromophenyl)-4-oxobutanoate

087054-1G 087054-2G 087054-5G
102852-220EA 490.77 USD
102852-220 102852-222 102852-224
Ethyl-4-(4-bromophenyl)-4-oxobutyrate ≥97%
Ethyl-4-(4-bromophenyl)-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₁₃BrO₃
MW: 285.14 g/mol
MDL Number: MFCD00672068
CAS Number: 30913-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR