4-(4-Chloro-2-methylphenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087052-1G 087052-2G 087052-5G
102852-208EA 490.77 USD
102852-208 102852-210 102852-212
4-(4-Chloro-2-methylphenyl)-4-oxobutyric acid ≥97%
4-(4-Chloro-2-methylphenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₁H₁₁ClO₃
MDL Number: MFCD09801804
CAS Number: 91193-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR