4-(2,4,6-Trimethylphenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087051-1G 087051-2G 087051-5G
102852-202EA 490.77 USD
102852-202 102852-204 102852-206
4-(2,4,6-Trimethylphenyl)-4-oxobutyric acid ≥97%
4-(2,4,6-Trimethylphenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₃H₁₆O₃
MDL Number: MFCD01096406
CAS Number: 15880-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR