4-(3,4,5-Trichlorophenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087046-1G 087046-2G 087046-5G
102852-172EA 476.92 USD
102852-172 102852-174 102852-176
4-(3,4,5-Trichlorophenyl)-4-oxobutyric acid ≥97%
4-(3,4,5-Trichlorophenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₀H₇Cl₃O₃
MDL Number: MFCD09801772
CAS Number: 62903-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR