4-(5-Chloro-2-methoxyphenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087044-1G 087044-2G 087044-5G
102852-160EA 490.77 USD
102852-160 102852-162 102852-164
4-(5-Chloro-2-methoxyphenyl)-4-oxobutyric acid ≥97%
4-(5-Chloro-2-methoxyphenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₁H₁₁ClO₄
MDL Number: MFCD07790541
CAS Number: 63213-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR