5-(4-t-Butylphenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

087042-1G 087042-2G 087042-5G
102852-148EA 592.31 USD
102852-148 102852-150 102852-152
5-(4-t-Butylphenyl)-5-oxovaleric acid ≥97%
5-(4-t-Butylphenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₅H₂₀O₃
MDL Number: MFCD02260962
CAS Number: 97692-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR