4-(2,5-Dichlorophenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

087041-1G 087041-2G 087041-5G
102852-142EA 592.31 USD
102852-142 102852-144 102852-146
4-(2,5-Dichlorophenyl)-4-oxobutyric acid ≥97%
4-(2,5-Dichlorophenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₀H₈Cl₂O₃
MDL Number: MFCD02260935
CAS Number: 52240-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR