4-(2,3-Dichlorophenyl)-4-oxobutyric acid ≥97%
Supplier: Matrix Scientific
087040-1G
087040-2G
087040-5G
102852-136EA
592.31
USD
102852-136
102852-138
102852-140
4-(2,3-Dichlorophenyl)-4-oxobutyric acid ≥97%
4-(2,3-Dichlorophenyl)-4-oxobutyric acid
Hazard: Irritant
Formula:
C₁₀H₈Cl₂O₃ |
MDL Number:
MFCD02260926 CAS Number: 32003-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...