5-Oxo-5-(3-trifluoromethylphenyl)valeric acid ≥97%

Supplier: Matrix Scientific

087039-1G 087039-2G 087039-5G
102852-130EA 592.31 USD
102852-130 102852-132 102852-134
5-Oxo-5-(3-trifluoromethylphenyl)valeric acid ≥97%
5-Oxo-5-(3-trifluoromethylphenyl)valeric acid

Hazard: Irritant

Formula: C₁₂H₁₁F₃O₃
MDL Number: MFCD02260917
CAS Number: 502650-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR