4-Oxo-4-(3-Trifluoromethylphenyl)butyric acid ≥97%

Supplier: Matrix Scientific

087038-1G 087038-2G 087038-5G
102852-124EA 592.31 USD
102852-124 102852-126 102852-128
4-Oxo-4-(3-Trifluoromethylphenyl)butyric acid ≥97%
4-Oxo-4-(3-Trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₁H₉F₃O₃
MDL Number: MFCD02260916
CAS Number: 56948-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR