7-Methyl-6-oxooctanoic acid ≥97%
Supplier: Matrix Scientific
087036-1G
087036-2G
087036-5G
102852-112EA
490.77
USD
102852-112
102852-114
102852-116
7-Methyl-6-oxooctanoic acid ≥97%
7-Methyl-6-oxooctanoic acid
Hazard: Irritant
Formula:
C₉H₁₆O₃ |
MDL Number:
MFCD02260881 CAS Number: 59210-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...