6-Methyl-5-oxoheptanoic acid ≥97%

Supplier: Matrix Scientific

087035-1G 087035-2G 087035-5G
102852-106EA 807.69 USD
102852-106 102852-108 102852-110
6-Methyl-5-oxoheptanoic acid ≥97%
6-Methyl-5-oxoheptanoic acid

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD02260880
CAS Number: 40564-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR