5-Methyl-4-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

087034-1G 087034-2G 087034-5G
102852-100EA 735.38 USD
102852-100 102852-102 102852-104
5-Methyl-4-oxohexanoic acid ≥97%
5-Methyl-4-oxohexanoic acid

Hazard: Irritant

Formula: C₇H₁₂O₃
MDL Number: MFCD00154360
CAS Number: 41654-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR