2-(1,3-Dioxolan-2-ylmethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087033-1G 087033-2G 087033-5G
102852-094EA 592.31 USD
102852-094 102852-096 102852-098
2-(1,3-Dioxolan-2-ylmethyl)benzoic acid ≥97%
2-(1,3-Dioxolan-2-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₄
MDL Number: MFCD07700149
CAS Number: 898767-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR