4-(2-Oxopropyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087030-1G 087030-2G 087030-5G
102852-076EA 886.15 USD
102852-076 102852-078 102852-080
4-(2-Oxopropyl)benzoic acid ≥97%
4-(2-Oxopropyl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD00238553
CAS Number: 15482-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR