2-(1-Oxo-2-phenylethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087023-1G 087023-2G 087023-5G
102852-034EA 735.38 USD
102852-034 102852-036 102852-038
2-(1-Oxo-2-phenylethyl)benzoic acid ≥97%
2-(1-Oxo-2-phenylethyl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂O₃
Melting Pt: 86…89 °C
MDL Number: MFCD00189277
CAS Number: 33148-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR