3-(1-Oxo-2-phenylethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087022-1G 087022-2G 087022-5G
102852-028EA 490.77 USD
102852-028 102852-030 102852-032
3-(1-Oxo-2-phenylethyl)benzoic acid ≥97%
3-(1-Oxo-2-phenylethyl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂O₃
MDL Number: MFCD01003946
CAS Number: 423151-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR