4-(1-Oxo-2-phenylethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

087021-1G 087021-2G 087021-5G
102852-022EA 490.77 USD
102852-022 102852-024 102852-026
4-(1-Oxo-2-phenylethyl)benzoic acid ≥97%
4-(1-Oxo-2-phenylethyl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂O₃
MDL Number: MFCD07700146
CAS Number: 108766-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR