4-Oxo-4-(4-pyridyl)butyric acid ≥97%
Supplier: Matrix Scientific
087020-1G
087020-2G
087020-5G
102852-016EA
735.38
USD
102852-016
102852-018
102852-020
4-Oxo-4-(4-pyridyl)butyric acid ≥97%
4-Oxo-4-(4-pyridyl)butyric acid
Hazard: Irritant
Formula:
C₉H₉NO₃ Melting Pt: 181…183 °C |
MDL Number:
MFCD01320178 CAS Number: 5693-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...