4-Oxo-4-(3-pyridyl)butyric acid ≥97%

Supplier: Matrix Scientific

087019-1G 087019-2G 087019-5G
102852-010EA 490.77 USD
102852-010 102852-012 102852-014
4-Oxo-4-(3-pyridyl)butyric acid ≥97%
4-Oxo-4-(3-pyridyl)butyric acid

Hazard: Irritant

Formula: C₉H₉NO₃
MDL Number: MFCD00870737
CAS Number: 4192-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR