4-Oxo-4-(2-pyridyl)butyric acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Oxo-4-(pyridin-2-yl)butanoic acid

087017-1G 087017-2G 087017-5G
102851-998EA 735.38 USD
102851-998 102852-000 102852-002
4-Oxo-4-(2-pyridyl)butyric acid ≥97%
4-Oxo-4-(2-pyridyl)butyric acid

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD01320175
CAS Number: 5768-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR