5-Oxo-6,6,6-trifluorohexanoic acid ≥97%

Supplier: Matrix Scientific

087016-1G 087016-2G 087016-5G
102851-992EA 886.15 USD
102851-992 102851-994 102851-996
5-Oxo-6,6,6-trifluorohexanoic acid ≥97%
5-Oxo-6,6,6-trifluorohexanoic acid

Hazard: Irritant

Formula: C₆H₇F₃O₃
Melting Pt: 43…45 °C
MDL Number: MFCD01320174
CAS Number: 184157-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR